The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178562.4 | NP_848657.1 | 3 - 4 | 551 | 70 |
XM_006715960.3 | XP_006716023.1 | 3 - 4 | 379 | 70 | |
GenBank mRNA | AF276891.1 | - | 3 - 4 | 286 | 70 |
AY236849.1 | - | 3 - 4 | 375 | 70 | |
BC044244.1 | - | 3 - 4 | 515 | 70 |