Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMARCC1
Entrez Gene ID
6599
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin subfamily c member 1
Gene Aliases
BAF155, CRACC1, Rsc8, SRG3, SWI3
Chromosome Location
Chr.3: 47585888 - 47781915 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003074.3 NP_003065.3 7 - 8 832 96
XM_011534034.1 XP_011532336.1 7 - 8 563 96
GenBank mRNA AK302541.1 - 7 - 8 607 96
BC021862.1 - 7 - 8 769 96
BC032564.1 - 7 - 8 816 96
BC039843.1 - 7 - 8 736 96
BC050564.1 - 7 - 8 761 96
BC065253.1 - 7 - 8 801 96
BC113465.1 - 7 - 8 780 96
BC117213.1 - 7 - 8 780 96
U66615.1 - 7 - 8 768 96

View Full Product Details