Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPO
Entrez Gene ID
130749
Gene Name
carboxypeptidase O
Gene Aliases
-
Chromosome Location
Chr.2: 206932853 - 206971064 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173077.2 NP_775100.1 7 - 8 821 73
XM_017003372.1 XP_016858861.1 8 - 9 6928 73
GenBank mRNA AJ422118.1 - 7 - 8 775 73
BC112076.1 - 7 - 8 821 73
BC112078.1 - 7 - 8 821 73

View Full Product Details