The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040428.3 | NP_001035518.1 | 7 - 8 | 1107 | 122 |
NM_018418.4 | NP_060888.2 | 8 - 9 | 1203 | 122 | |
XM_005267851.1 | XP_005267908.1 | 8 - 9 | 1177 | 122 | |
XM_005267852.1 | XP_005267909.1 | 7 - 8 | 1106 | 122 | |
XM_005267854.1 | XP_005267911.1 | 9 - 10 | 1239 | 122 | |
XM_005267855.1 | XP_005267912.1 | 8 - 9 | 1172 | 122 | |
XM_006720204.1 | XP_006720267.1 | 8 - 9 | 1177 | 122 | |
XM_006720205.1 | XP_006720268.1 | 8 - 9 | 1177 | 122 | |
XM_011536951.1 | XP_011535253.1 | 8 - 9 | 1371 | 122 | |
XM_011536952.1 | XP_011535254.1 | 6 - 7 | 989 | 122 | |
XM_011536953.1 | XP_011535255.1 | 7 - 8 | 1310 | 122 | |
XM_017021452.1 | XP_016876941.1 | 8 - 9 | 1368 | 122 | |
XM_017021453.1 | XP_016876942.1 | 8 - 9 | 1337 | 122 | |
XM_017021454.1 | XP_016876943.1 | 9 - 10 | 1236 | 122 | |
XM_017021455.1 | XP_016876944.1 | 8 - 9 | 1169 | 122 | |
XM_017021456.1 | XP_016876945.1 | 9 - 10 | 1182 | 122 | |
XM_017021457.1 | XP_016876946.1 | 7 - 8 | 1307 | 122 | |
GenBank mRNA | AF144487.3 | - | 8 - 9 | 1183 | 122 |
AF144488.2 | - | 6 - 7 | 911 | 122 | |
AK055864.1 | - | 7 - 8 | 2775 | 122 | |
AK225746.1 | - | 9 - 10 | 1177 | 122 | |
AK290631.1 | - | 7 - 8 | 1078 | 122 | |
AL136604.1 | - | 7 - 8 | 1087 | 122 | |
BC008656.1 | - | 3 - 4 | 635 | 122 | |
BC090875.1 | - | 8 - 9 | 1490 | 122 | |
CR533563.1 | - | 7 - 8 | 932 | 122 | |
CR936777.1 | - | 8 - 9 | 1149 | 122 | |
EU668353.1 | - | 8 - 9 | 1179 | 122 |