The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_174922.4 | NP_777582.4 | 4 - 5 | 426 | 66 |
XM_011516907.2 | XP_011515209.1 | 4 - 5 | 1139 | 66 | |
XM_011516909.1 | XP_011515211.1 | 4 - 5 | 426 | 66 | |
XM_017013174.1 | XP_016868663.1 | 4 - 5 | 464 | 66 | |
GenBank mRNA | AK092773.1 | - | 4 - 5 | 389 | 66 |
BC101659.1 | - | 4 - 5 | 366 | 66 | |
BC143326.1 | - | - | 366 | 66 |