Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACTR3
Entrez Gene ID
10096
Gene Name
ARP3 actin related protein 3 homolog
Gene Aliases
ARP3
Chromosome Location
Chr.2: 113889934 - 113961554 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277140.1 NP_001264069.1 7 - 8 815 127
NM_005721.4 NP_005712.1 7 - 8 1024 127
NR_102318.1 6 - 7 899 127
GenBank mRNA AB209353.1 - 5 - 6 533 127
AF006083.1 - 7 - 8 850 127
AF127773.1 - 7 - 8 893 127
AK300030.1 - 6 - 7 723 127
AK300224.1 - 6 - 7 645 127
AK302152.1 - 7 - 8 769 127
AK302359.1 - 7 - 8 803 127
AK312659.1 - 7 - 8 824 127
AK316213.1 - 7 - 8 846 127
AK316396.1 - 7 - 8 815 127
BC044590.1 - 7 - 8 998 127

View Full Product Details