The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168474.1 | NP_001161946.1 | 2 - 3 | 181 | 96 |
NM_024885.3 | NP_079161.3 | 2 - 3 | 349 | 96 | |
XM_005262152.1 | XP_005262209.1 | 2 - 3 | 181 | 96 | |
XM_006724664.1 | XP_006724727.1 | 2 - 3 | 348 | 96 | |
GenBank mRNA | AF285595.1 | - | 2 - 3 | 183 | 96 |
AK026810.1 | - | 1 - 2 | 327 | 96 | |
AK308261.1 | - | 2 - 3 | 350 | 96 | |
BC043391.1 | - | 2 - 3 | 335 | 96 | |
DB042169.1 | - | 2 - 3 | 181 | 96 |