Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TAF7L
Entrez Gene ID
54457
Gene Name
TATA-box binding protein associated factor 7 like
Gene Aliases
CT40, TAF2Q
Chromosome Location
Chr.X: 101268253 - 101293071 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168474.1 NP_001161946.1 4 - 5 394 132
NM_024885.3 NP_079161.3 4 - 5 562 132
XM_005262152.1 XP_005262209.1 4 - 5 394 132
XM_006724664.1 XP_006724727.1 4 - 5 561 132
GenBank mRNA AF285595.1 - 4 - 5 396 132
AK026810.1 - 3 - 4 540 132
AK308261.1 - 4 - 5 563 132
BC043391.1 - 4 - 5 548 132
DB042169.1 - 4 - 5 394 132

View Full Product Details