Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TAF7L
Entrez Gene ID
54457
Gene Name
TATA-box binding protein associated factor 7 like
Gene Aliases
CT40, TAF2Q
Chromosome Location
Chr.X: 101268253 - 101293071 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168474.1 NP_001161946.1 11 - 12 1140 77
NM_024885.3 NP_079161.3 11 - 12 1308 77
XM_005262152.1 XP_005262209.1 11 - 12 1143 77
XM_006724664.1 XP_006724727.1 11 - 12 1310 77
GenBank mRNA AF285595.1 - 11 - 12 1136 77
AK026810.1 - 9 - 10 1064 77
AK308261.1 - 11 - 12 1310 77
BC043391.1 - 11 - 12 1294 77

View Full Product Details