The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001174116.1 | NP_001167587.1 | 40 - 41 | 8855 | 87 |
NM_001174117.1 | NP_001167588.1 | 38 - 39 | 6944 | 87 | |
NM_015263.3 | NP_056078.2 | 40 - 41 | 8852 | 87 | |
XM_005254255.1 | XP_005254312.1 | 41 - 42 | 8981 | 87 | |
XM_005254256.1 | XP_005254313.1 | 41 - 42 | 8978 | 87 | |
XM_017022034.1 | XP_016877523.1 | 41 - 42 | 8654 | 87 | |
GenBank mRNA | AB020663.2 | - | 28 - 29 | 6232 | 87 |
AF389880.1 | - | 40 - 41 | 8834 | 87 | |
BC016032.1 | - | 18 - 19 | 3067 | 87 | |
BC022050.1 | - | 11 - 12 | 1172 | 87 | |
BC140781.1 | - | 38 - 39 | 6809 | 87 | |
BC144539.1 | - | 40 - 41 | 8720 | 87 |