The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173582.4 | NP_775853.2 | 4 - 5 | 774 | 86 |
XM_011544953.2 | XP_011543255.1 | 5 - 6 | 829 | 86 | |
GenBank mRNA | AB019210.1 | - | 4 - 5 | 725 | 86 |
AK056591.1 | - | 4 - 5 | 732 | 86 | |
BC059360.1 | - | 4 - 5 | 738 | 86 |