Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AIF1L
Entrez Gene ID
83543
Gene Name
allograft inflammatory factor 1 like
Gene Aliases
C9orf58, IBA2
Chromosome Location
Chr.9: 131096476 - 131123152 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001185095.1 NP_001172024.1 1 - 2 181 61
NM_001185096.1 NP_001172025.1 1 - 2 181 61
NM_031426.3 NP_113614.1 1 - 2 181 61
GenBank mRNA AK022845.1 - 1 - 2 126 61
AK091912.1 - 1 - 2 118 61
AK128526.1 - 1 - 2 126 61
AK314600.1 - 1 - 2 128 61
AL136566.1 - 1 - 2 115 61
AL833896.1 - 1 - 2 115 61
AY359067.1 - 1 - 2 132 61
BC021253.2 - 1 - 2 106 61
DA285481.1 - 1 - 2 182 61

View Full Product Details