Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CSNK2B
Entrez Gene ID
1460
Gene Name
casein kinase 2 beta
Gene Aliases
CK2B, CK2N, CSK2B, G5A
Chromosome Location
Chr.6: 31665880 - 31670070 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320.6 NP_001311.3 5 - 6 709 59
GenBank mRNA AK311860.1 - 5 - 6 488 59
AY113186.1 - 4 - 5 369 59
BC112017.1 - 5 - 6 522 59
BC112019.1 - 5 - 6 522 59
CR541699.1 - 4 - 5 369 59
DB570461.2 - 3 - 4 271 59
HE864446.1 - 4 - 5 381 59
X16312.1 - 5 - 6 502 59
X16937.1 - 5 - 6 502 59

View Full Product Details