Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
METTL17
Entrez Gene ID
64745
Gene Name
methyltransferase like 17
Gene Aliases
METT11D1
Chromosome Location
Chr.14: 20989806 - 20997035 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029991.1 NP_001025162.1 3 - 4 558 70
NM_022734.2 NP_073571.1 3 - 4 558 70
GenBank mRNA AF321002.1 - 3 - 4 393 70
AK024512.1 - 3 - 4 384 70
BC005053.1 - 3 - 4 377 70
KU178747.1 - 3 - 4 361 70
KU178749.1 - 3 - 4 361 70

View Full Product Details