The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001029991.1 | NP_001025162.1 | 9 - 10 | 1072 | 81 |
NM_022734.2 | NP_073571.1 | 9 - 10 | 1072 | 81 | |
XM_006720236.2 | XP_006720299.1 | 8 - 9 | 832 | 81 | |
XM_017021601.1 | XP_016877090.1 | 8 - 9 | 832 | 81 | |
GenBank mRNA | AF321002.1 | - | 9 - 10 | 907 | 81 |
AK024512.1 | - | 9 - 10 | 898 | 81 | |
BC005053.1 | - | 9 - 10 | 891 | 81 | |
KU178747.1 | - | 9 - 10 | 875 | 81 | |
KU178748.1 | - | 9 - 10 | 907 | 81 | |
KU178749.1 | - | 9 - 10 | 875 | 81 |