Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRM1
Entrez Gene ID
79922
Gene Name
mitochondrial rRNA methyltransferase 1
Gene Aliases
-
Chromosome Location
Chr.17: 36601581 - 36634698 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024864.4 NP_079140.2 2 - 3 856 132
XM_011525275.2 XP_011523577.1 2 - 3 871 132
XM_011525276.2 XP_011523578.1 2 - 3 871 132
XM_017025112.1 XP_016880601.1 2 - 3 871 132
GenBank mRNA AK026231.1 - 2 - 3 314 132
AK225449.1 - 2 - 3 314 132
AK292307.1 - 2 - 3 862 132
BC009416.1 - 2 - 3 357 132
BC072411.1 - 2 - 3 796 132
CR457357.1 - 1 - 2 47 132

View Full Product Details