The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256827.1 | NP_001243756.1 | 3 - 4 | 435 | 75 |
NM_001256828.1 | NP_001243757.1 | 2 - 3 | 192 | 75 | |
NM_017575.4 | NP_060045.4 | 10 - 11 | 2919 | 75 | |
XM_005256569.3 | XP_005256626.1 | 10 - 11 | 3117 | 75 | |
XM_005256571.4 | XP_005256628.1 | 2 - 3 | 1502 | 75 | |
XM_011523769.2 | XP_011522071.1 | 11 - 12 | 3107 | 75 | |
XM_011523775.2 | XP_011522077.1 | 3 - 4 | 1511 | 75 | |
XM_017024399.1 | XP_016879888.1 | 10 - 11 | 1977 | 75 | |
GenBank mRNA | AB018275.2 | - | 10 - 11 | 2956 | 75 |
AK302964.1 | - | 2 - 3 | 192 | 75 | |
AY145883.1 | - | 10 - 11 | 2869 | 75 | |
AY168921.1 | - | 9 - 10 | 2771 | 75 | |
BC064916.1 | - | 3 - 4 | 435 | 75 |