The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006612.5 | NP_006603.2 | 4 - 5 | 540 | 75 |
XM_005256424.2 | XP_005256481.1 | 5 - 6 | 531 | 75 | |
GenBank mRNA | AB014606.2 | - | 4 - 5 | 536 | 75 |
AK307401.1 | - | 4 - 5 | 383 | 75 | |
BC034993.1 | - | 4 - 5 | 514 | 75 | |
BM458656.1 | - | 4 - 5 | 540 | 75 | |
U91329.1 | - | 3 - 4 | 296 | 75 |