The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033281.5 | NP_150597.1 | 2 - 3 | 192 | 67 |
GenBank mRNA | AF271777.1 | - | - | 296 | 67 |
BC015966.1 | - | 2 - 3 | 182 | 67 | |
BC017900.1 | - | 2 - 3 | 122 | 67 | |
BX951861.1 | - | 2 - 3 | 198 | 67 |