The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077199.2 | NP_001070667.1 | 11 - 12 | 1889 | 78 |
NM_001270492.1 | NP_001257421.1 | 12 - 13 | 2010 | 78 | |
NM_001323527.1 | NP_001310456.1 | 12 - 13 | 2013 | 78 | |
NM_001323529.1 | NP_001310458.1 | 11 - 12 | 1886 | 78 | |
NM_001323534.1 | NP_001310463.1 | 12 - 13 | 1904 | 78 | |
NM_001323535.1 | NP_001310464.1 | 11 - 12 | 1777 | 78 | |
NM_139168.3 | NP_631907.1 | 12 - 13 | 2042 | 78 | |
XM_011543171.2 | XP_011541473.1 | 11 - 12 | 2114 | 78 | |
XM_017009055.1 | XP_016864544.1 | 10 - 11 | 1423 | 78 | |
XM_017009056.1 | XP_016864545.1 | 11 - 12 | 1674 | 78 | |
GenBank mRNA | AF459094.1 | - | 11 - 12 | 1723 | 78 |
AK091758.1 | - | 11 - 12 | 1771 | 78 | |
AK125893.1 | - | 4 - 5 | 1574 | 78 | |
AK308287.1 | - | 12 - 13 | 1912 | 78 | |
BC067770.1 | - | 12 - 13 | 2043 | 78 | |
BC112343.1 | - | 9 - 10 | 1411 | 78 |