Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DCX
Entrez Gene ID
1641
Gene Name
doublecortin
Gene Aliases
DBCN, DC, LISX, SCLH, XLIS
Chromosome Location
Chr.X: 111293779 - 111412232 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000555.3 NP_000546.2 1 - 2 388 145
GenBank mRNA AJ003112.1 - 1 - 2 388 145
AK297297.1 - 1 - 2 191 145

View Full Product Details