Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SF3B3
Entrez Gene ID
23450
Gene Name
splicing factor 3b subunit 3
Gene Aliases
RSE1, SAP130, SF3b130, STAF130
Chromosome Location
Chr.16: 70523788 - 70577668 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012426.4 NP_036558.3 13 - 14 1920 67
GenBank mRNA AJ001443.1 - 13 - 14 1946 67
AK001297.1 - 2 - 3 299 67
AK074933.1 - 7 - 8 815 67
AK291768.1 - 13 - 14 1918 67
BC068974.1 - 13 - 14 1857 67
D87686.1 - 13 - 14 1865 67

View Full Product Details