Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SF3B3
Entrez Gene ID
23450
Gene Name
splicing factor 3b subunit 3
Gene Aliases
RSE1, SAP130, SF3b130, STAF130
Chromosome Location
Chr.16: 70523788 - 70577668 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012426.4 NP_036558.3 17 - 18 2502 62
GenBank mRNA AJ001443.1 - 17 - 18 2528 62
AK001297.1 - 6 - 7 881 62
AK074933.1 - 11 - 12 1397 62
AK291768.1 - 17 - 18 2500 62
BC068974.1 - 17 - 18 2439 62
D87686.1 - 17 - 18 2447 62

View Full Product Details