The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012426.4 | NP_036558.3 | 25 - 26 | 3725 | 76 |
GenBank mRNA | AJ001443.1 | - | 25 - 26 | 3751 | 76 |
AK001297.1 | - | 14 - 15 | 2104 | 76 | |
AK074933.1 | - | 19 - 20 | 2620 | 76 | |
AK291768.1 | - | 25 - 26 | 3723 | 76 | |
AL110251.1 | - | 4 - 5 | 509 | 76 | |
BC000463.1 | - | 7 - 8 | 1069 | 76 | |
BC003146.1 | - | 7 - 8 | 1074 | 76 | |
BC068974.1 | - | 25 - 26 | 3662 | 76 | |
D13642.1 | - | 8 - 9 | 1196 | 76 | |
D87686.1 | - | 25 - 26 | 3670 | 76 |