Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SF3B3
Entrez Gene ID
23450
Gene Name
splicing factor 3b subunit 3
Gene Aliases
RSE1, SAP130, SF3b130, STAF130
Chromosome Location
Chr.16: 70523788 - 70577668 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012426.4 NP_036558.3 8 - 9 1276 116
GenBank mRNA AJ001443.1 - 8 - 9 1302 116
AK074933.1 - 2 - 3 171 116
AK291768.1 - 8 - 9 1274 116
BC068974.1 - 8 - 9 1213 116
D87686.1 - 8 - 9 1221 116

View Full Product Details