The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114380.1 | NP_001107852.1 | 24 - 25 | 2901 | 115 |
NM_002209.2 | NP_002200.2 | 26 - 27 | 3153 | 115 | |
XM_005255313.1 | XP_005255370.1 | 26 - 27 | 3070 | 115 | |
XM_006721044.1 | XP_006721107.1 | 24 - 25 | 2855 | 115 | |
XM_011545849.1 | XP_011544151.1 | 26 - 27 | 3086 | 115 | |
GenBank mRNA | AK303190.1 | - | 8 - 9 | 766 | 115 |
AK314246.1 | - | 26 - 27 | 3107 | 115 | |
BC008777.2 | - | 24 - 25 | 2834 | 115 | |
Y00796.1 | - | 26 - 27 | 3065 | 115 |