The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114380.1 | NP_001107852.1 | 26 - 27 | 3062 | 83 |
NM_002209.2 | NP_002200.2 | 28 - 29 | 3314 | 83 | |
XM_005255313.1 | XP_005255370.1 | 28 - 29 | 3231 | 83 | |
XM_006721044.1 | XP_006721107.1 | 26 - 27 | 3016 | 83 | |
XM_011545849.1 | XP_011544151.1 | 28 - 29 | 3247 | 83 | |
GenBank mRNA | AK303190.1 | - | 10 - 11 | 927 | 83 |
AK314246.1 | - | 28 - 29 | 3268 | 83 | |
BC008777.2 | - | 26 - 27 | 2995 | 83 | |
Y00796.1 | - | 28 - 29 | 3226 | 83 |