Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NPSR1
Entrez Gene ID
387129
Gene Name
neuropeptide S receptor 1
Gene Aliases
ASRT2, GPR154, GPRA, NPSR, PGR14, VRR1
Chromosome Location
Chr.7: 34658239 - 34878332 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300933.1 NP_001287862.1 4 - 5 621 68
NM_001300935.1 NP_001287864.1 4 - 5 654 68
NM_207172.1 NP_997055.1 4 - 5 608 68
NM_207173.1 NP_997056.1 4 - 5 608 68
GenBank mRNA AY310326.1 - 4 - 5 608 68
AY310327.1 - 4 - 5 608 68
AY310328.1 - 4 - 5 575 68
AY310330.1 - 3 - 4 504 68
AY956446.1 - 4 - 5 513 68
AY956447.1 - 4 - 5 484 68
AY956448.1 - 4 - 5 484 68
BC132693.1 - 4 - 5 525 68
BC132695.1 - 4 - 5 525 68

View Full Product Details