The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004327.3 | NP_004318.3 | 6 - 7 | 2516 | 58 |
NM_021574.2 | NP_067585.2 | 6 - 7 | 2516 | 58 | |
GenBank mRNA | AB209991.1 | - | 6 - 7 | 2265 | 58 |
AB742171.1 | - | 2 - 3 | 233 | 58 | |
AK303241.1 | - | 6 - 7 | 950 | 58 | |
AY533677.1 | - | 4 - 5 | 372 | 58 | |
BC066122.1 | - | 6 - 7 | 2347 | 58 | |
X02596.1 | - | 6 - 7 | 2408 | 58 | |
Y00661.1 | - | 6 - 7 | 2516 | 58 |