The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004327.3 | NP_004318.3 | 7 - 8 | 2569 | 65 |
NM_021574.2 | NP_067585.2 | 7 - 8 | 2569 | 65 | |
GenBank mRNA | AB209991.1 | - | 7 - 8 | 2318 | 65 |
AK303241.1 | - | 7 - 8 | 1003 | 65 | |
AY533677.1 | - | 5 - 6 | 425 | 65 | |
BC066122.1 | - | 7 - 8 | 2400 | 65 | |
X02596.1 | - | 7 - 8 | 2461 | 65 | |
Y00661.1 | - | 7 - 8 | 2569 | 65 |