The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000059.3 | NP_000050.2 | 21 - 22 | 8982 | 136 |
GenBank mRNA | U43746.1 | - | 21 - 22 | 8983 | 136 |