The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_170692.2 | NP_733793.2 | 1 - 2 | 965 | 82 |
XM_011510166.2 | XP_011508468.1 | 1 - 2 | 965 | 82 | |
XM_011510167.2 | XP_011508469.1 | 1 - 2 | 965 | 82 | |
GenBank mRNA | AB209789.1 | - | 1 - 2 | 873 | 82 |
AK075169.1 | - | 1 - 2 | 965 | 82 | |
AK311017.1 | - | 1 - 2 | 964 | 82 | |
AV706305.1 | - | 1 - 2 | 252 | 82 | |
BC041045.1 | - | - | 518 | 82 | |
BC063442.1 | - | 1 - 2 | 525 | 82 | |
BC078167.1 | - | 1 - 2 | 587 | 82 | |
BC110611.2 | - | 1 - 2 | 197 | 82 |