The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318785.1 | NP_001305714.1 | 3 - 4 | 504 | 105 |
NM_020320.4 | NP_064716.2 | 3 - 4 | 283 | 105 | |
NR_134857.1 | 3 - 4 | 283 | 105 | ||
XM_011535948.2 | XP_011534250.1 | 3 - 4 | 268 | 105 | |
XM_011535949.2 | XP_011534251.1 | 3 - 4 | 268 | 105 | |
XM_017011072.1 | XP_016866561.1 | 3 - 4 | 449 | 105 | |
XM_017011073.1 | XP_016866562.1 | 4 - 5 | 667 | 105 | |
XM_017011074.1 | XP_016866563.1 | 3 - 4 | 270 | 105 | |
XM_017011075.1 | XP_016866564.1 | 3 - 4 | 449 | 105 | |
XM_017011076.1 | XP_016866565.1 | 3 - 4 | 449 | 105 | |
XM_017011077.1 | XP_016866566.1 | 3 - 4 | 226 | 105 | |
XM_017011078.1 | XP_016866567.1 | 3 - 4 | 270 | 105 | |
XM_017011079.1 | XP_016866568.1 | 3 - 4 | 449 | 105 | |
XM_017011080.1 | XP_016866569.1 | 3 - 4 | 449 | 105 | |
XM_017011081.1 | XP_016866570.1 | 3 - 4 | 270 | 105 | |
GenBank mRNA | AK023550.1 | - | 3 - 4 | 226 | 105 |
AK093934.1 | - | 3 - 4 | 656 | 105 | |
AK315669.1 | - | 3 - 4 | 220 | 105 | |
BC010420.1 | - | 3 - 4 | 254 | 105 | |
BC105784.1 | - | 3 - 4 | 443 | 105 | |
DA607375.1 | - | 3 - 4 | 269 | 105 | |
DB455377.1 | - | 3 - 4 | 285 | 105 |