The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001671.3 | NP_001001671.3 | 11 - 12 | 1691 | 80 |
XM_011545507.2 | XP_011543809.2 | 12 - 13 | 1918 | 80 | |
XM_011545508.2 | XP_011543810.2 | 12 - 13 | 1918 | 80 | |
XM_011545510.2 | XP_011543812.1 | 5 - 6 | 2431 | 80 | |
XM_011545511.2 | XP_011543813.1 | 2 - 3 | 942 | 80 | |
XM_017029511.1 | XP_016885000.1 | 12 - 13 | 1920 | 80 | |
GenBank mRNA | AK131412.1 | - | 8 - 9 | 1019 | 80 |
AK131477.1 | - | 11 - 12 | 1653 | 80 |