The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001671.3 | NP_001001671.3 | 14 - 15 | 1925 | 76 |
XM_011545507.2 | XP_011543809.2 | 15 - 16 | 2152 | 76 | |
XM_011545508.2 | XP_011543810.2 | 15 - 16 | 2152 | 76 | |
XM_011545510.2 | XP_011543812.1 | 8 - 9 | 2665 | 76 | |
XM_011545511.2 | XP_011543813.1 | 5 - 6 | 1176 | 76 | |
XM_017029511.1 | XP_016885000.1 | 15 - 16 | 2154 | 76 | |
GenBank mRNA | AK131412.1 | - | 11 - 12 | 1253 | 76 |
AK131477.1 | - | 15 - 16 | 1985 | 76 |