The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001671.3 | NP_001001671.3 | 6 - 7 | 992 | 96 |
XM_011545507.2 | XP_011543809.2 | 7 - 8 | 1219 | 96 | |
XM_011545508.2 | XP_011543810.2 | 7 - 8 | 1219 | 96 | |
XM_017029511.1 | XP_016885000.1 | 7 - 8 | 1221 | 96 | |
GenBank mRNA | AK131412.1 | - | 2 - 3 | 247 | 96 |
AK131477.1 | - | 5 - 6 | 881 | 96 |