The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153221.2 | NP_694953.2 | 2 - 3 | 250 | 128 |
GenBank mRNA | AF542080.1 | - | 2 - 3 | 218 | 128 |
AK314213.1 | - | 2 - 3 | 222 | 128 | |
DA328640.1 | - | 2 - 3 | 250 | 128 |