The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025245.2 | NP_079521.1 | 4 - 5 | 920 | 80 |
NR_038198.1 | 4 - 5 | 819 | 80 | ||
XM_006722911.3 | XP_006722974.1 | 3 - 4 | 501 | 80 | |
XM_011528320.2 | XP_011526622.1 | 5 - 6 | 966 | 80 | |
XM_011528322.2 | XP_011526624.1 | 3 - 4 | 555 | 80 | |
XM_017027326.1 | XP_016882815.1 | 5 - 6 | 601 | 80 | |
XM_017027327.1 | XP_016882816.1 | 5 - 6 | 582 | 80 | |
XM_017027328.1 | XP_016882817.1 | 5 - 6 | 966 | 80 | |
XM_017027329.1 | XP_016882818.1 | 2 - 3 | 391 | 80 | |
GenBank mRNA | AJ300182.1 | - | 3 - 4 | 501 | 80 |
AK097427.1 | - | 4 - 5 | 657 | 80 | |
AK301631.1 | - | 4 - 5 | 584 | 80 | |
BC141859.1 | - | 4 - 5 | 920 | 80 | |
BC143588.1 | - | 4 - 5 | 595 | 80 |