Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LECT2
Entrez Gene ID
3950
Gene Name
leukocyte cell derived chemotaxin 2
Gene Aliases
chm-II, chm2
Chromosome Location
Chr.5: 135946911 - 135955034 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002302.2 NP_002293.2 2 - 3 344 82
GenBank mRNA AK314092.1 - 2 - 3 293 82
AV661075.1 - 2 - 3 292 82
BC093670.1 - 2 - 3 205 82
BC101579.1 - 2 - 3 205 82
BI825931.1 - 1 - 2 266 81
D63521.1 - 2 - 3 344 82

View Full Product Details