The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022734.2 | NP_073571.1 | 13 - 14 | 1464 | 74 |
XM_017021601.1 | XP_016877090.1 | 12 - 13 | 1224 | 74 | |
GenBank mRNA | AK024512.1 | - | 13 - 14 | 1290 | 74 |
KU178749.1 | - | 13 - 14 | 1267 | 74 |