The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014586.1 | NP_055401.1 | 10 - 11 | 1842 | 74 |
XM_011529537.2 | XP_011527839.1 | 9 - 10 | 1458 | 74 | |
GenBank mRNA | AJ271722.1 | - | 10 - 11 | 1842 | 74 |