The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145076.2 | NP_001138548.1 | 18 - 19 | 2251 | 117 |
NM_019063.4 | NP_061936.2 | 19 - 20 | 2425 | 117 | |
XM_005264267.1 | XP_005264324.1 | 20 - 21 | 2438 | 117 | |
XM_005264268.3 | XP_005264325.1 | 20 - 21 | 2438 | 117 | |
XM_006711991.1 | XP_006712054.1 | 19 - 20 | 2264 | 117 | |
XM_006711992.2 | XP_006712055.1 | 21 - 22 | 2559 | 117 | |
GenBank mRNA | AF177377.2 | - | 19 - 20 | 2387 | 117 |
AK056880.1 | - | 8 - 9 | 927 | 117 | |
AK314702.1 | - | 19 - 20 | 2399 | 117 | |
AL832392.1 | - | 5 - 6 | 578 | 117 | |
BC140845.1 | - | 19 - 20 | 2348 | 117 | |
BC146799.1 | - | 18 - 19 | 2225 | 117 | |
BX647731.1 | - | 10 - 11 | 1488 | 117 |