The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145076.2 | NP_001138548.1 | 20 - 21 | 2447 | 85 |
NM_019063.4 | NP_061936.2 | 21 - 22 | 2621 | 85 | |
XM_005264267.1 | XP_005264324.1 | 22 - 23 | 2634 | 85 | |
XM_006711991.1 | XP_006712054.1 | 21 - 22 | 2460 | 85 | |
XM_006711992.2 | XP_006712055.1 | 23 - 24 | 2755 | 85 | |
GenBank mRNA | AF177377.2 | - | 21 - 22 | 2583 | 85 |
AK056880.1 | - | 10 - 11 | 1123 | 85 | |
AK295778.1 | - | 6 - 7 | 659 | 85 | |
AK314702.1 | - | 21 - 22 | 2595 | 85 | |
AL832392.1 | - | 7 - 8 | 774 | 85 | |
BC140845.1 | - | 21 - 22 | 2544 | 85 | |
BC146799.1 | - | 20 - 21 | 2421 | 85 | |
BX647731.1 | - | 12 - 13 | 1684 | 85 |