Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EML4
Entrez Gene ID
27436
Gene Name
echinoderm microtubule associated protein like 4
Gene Aliases
C2orf2, ELP120, EMAP-4, EMAPL4, ROPP120
Chromosome Location
Chr.2: 42169338 - 42332548 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145076.2 NP_001138548.1 2 - 3 484 106
NM_019063.4 NP_061936.2 2 - 3 484 106
XM_005264267.1 XP_005264324.1 2 - 3 464 106
XM_005264268.3 XP_005264325.1 2 - 3 464 106
XM_006711991.1 XP_006712054.1 2 - 3 464 106
XM_006711992.2 XP_006712055.1 3 - 4 585 106
GenBank mRNA AF177377.2 - 2 - 3 446 106
AK001804.1 - - 1527 106
AK314702.1 - 2 - 3 458 106
BC104647.1 - 2 - 3 411 106
BC140845.1 - 2 - 3 407 106
BC146799.1 - 2 - 3 458 106

View Full Product Details