The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013371.3 | NP_037503.2 | 3 - 4 | 1078 | 89 |
NM_153758.2 | NP_715639.1 | 2 - 3 | 278 | 89 | |
XM_011509450.2 | XP_011507752.1 | 2 - 3 | 717 | 89 | |
GenBank mRNA | AF192498.1 | - | 2 - 3 | 186 | 89 |
AF453946.1 | - | 3 - 4 | 1078 | 89 | |
AY040367.1 | - | 2 - 3 | 278 | 89 | |
FJ380053.1 | - | 1 - 2 | 139 | 89 |