The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286455.1 | NP_001273384.1 | 4 - 5 | 623 | 69 |
NM_015482.1 | NP_056297.1 | 4 - 5 | 1082 | 69 | |
NM_021945.6 | NP_068764.3 | 5 - 6 | 868 | 69 | |
XM_011514801.2 | XP_011513103.1 | 5 - 6 | 1169 | 69 | |
XM_017011180.1 | XP_016866669.1 | 5 - 6 | 1646 | 69 | |
XM_017011181.1 | XP_016866670.1 | 3 - 4 | 395 | 69 | |
XM_017011182.1 | XP_016866671.1 | 2 - 3 | 677 | 69 | |
XM_017011184.1 | XP_016866673.1 | 2 - 3 | 332 | 69 | |
XM_017011186.1 | XP_016866675.1 | 5 - 6 | 565 | 69 | |
GenBank mRNA | AK172770.1 | - | 1 - 2 | 178 | 69 |
AK308282.1 | - | 5 - 6 | 622 | 69 | |
AK308943.1 | - | 4 - 5 | 566 | 69 | |
BC038748.1 | - | 5 - 6 | 839 | 69 | |
BC128580.1 | - | 4 - 5 | 575 | 69 |