The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286455.1 | NP_001273384.1 | 5 - 6 | 752 | 59 |
NM_015482.1 | NP_056297.1 | 5 - 6 | 1211 | 59 | |
NM_021945.6 | NP_068764.3 | 6 - 7 | 997 | 59 | |
NR_104448.1 | 6 - 7 | 805 | 59 | ||
XM_011514801.2 | XP_011513103.1 | 6 - 7 | 1298 | 59 | |
XM_017011180.1 | XP_016866669.1 | 6 - 7 | 1775 | 59 | |
XM_017011181.1 | XP_016866670.1 | 4 - 5 | 524 | 59 | |
XM_017011182.1 | XP_016866671.1 | 3 - 4 | 806 | 59 | |
XM_017011184.1 | XP_016866673.1 | 3 - 4 | 461 | 59 | |
XM_017011186.1 | XP_016866675.1 | 6 - 7 | 694 | 59 | |
GenBank mRNA | AK172770.1 | - | 2 - 3 | 307 | 59 |
AK308282.1 | - | 6 - 7 | 751 | 59 | |
AK308943.1 | - | 5 - 6 | 695 | 59 | |
BC038748.1 | - | 6 - 7 | 968 | 59 | |
BC128580.1 | - | 5 - 6 | 704 | 59 | |
BC128581.1 | - | 6 - 7 | 761 | 59 |