The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286455.1 | NP_001273384.1 | 6 - 7 | 856 | 66 |
NM_015482.1 | NP_056297.1 | 6 - 7 | 1315 | 66 | |
NM_021945.6 | NP_068764.3 | 7 - 8 | 1101 | 66 | |
NR_104448.1 | 7 - 8 | 909 | 66 | ||
XM_011514801.2 | XP_011513103.1 | 7 - 8 | 1402 | 66 | |
XM_017011180.1 | XP_016866669.1 | 7 - 8 | 1879 | 66 | |
XM_017011181.1 | XP_016866670.1 | 5 - 6 | 628 | 66 | |
XM_017011182.1 | XP_016866671.1 | 4 - 5 | 910 | 66 | |
XM_017011184.1 | XP_016866673.1 | 4 - 5 | 565 | 66 | |
XM_017011186.1 | XP_016866675.1 | 7 - 8 | 798 | 66 | |
GenBank mRNA | AK172770.1 | - | 3 - 4 | 411 | 66 |
AK308282.1 | - | 7 - 8 | 855 | 66 | |
AK308943.1 | - | 6 - 7 | 799 | 66 | |
BC038748.1 | - | 7 - 8 | 1072 | 66 | |
BC128580.1 | - | 6 - 7 | 808 | 66 | |
BC128581.1 | - | 7 - 8 | 865 | 66 |