The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001261392.1 | NP_001248321.1 | 12 - 13 | 1070 | 113 |
NM_001261394.1 | NP_001248323.1 | 12 - 13 | 998 | 113 | |
NM_024894.3 | NP_079170.2 | 13 - 14 | 1148 | 113 | |
NR_048552.1 | 12 - 13 | 1102 | 113 | ||
XM_011510398.2 | XP_011508700.1 | 12 - 13 | 1015 | 113 | |
XM_011510399.2 | XP_011508701.1 | 11 - 12 | 953 | 113 | |
GenBank mRNA | AK024137.1 | - | 12 - 13 | 983 | 113 |
AK290680.1 | - | 13 - 14 | 1112 | 113 | |
AK297169.1 | - | 12 - 13 | 1058 | 113 | |
AK299617.1 | - | 12 - 13 | 1090 | 113 | |
BC005125.2 | - | 13 - 14 | 1128 | 113 |