The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001294345.1 | NP_001281274.1 | 3 - 4 | 480 | 70 |
NM_001294346.1 | NP_001281275.1 | 3 - 4 | 587 | 70 | |
NM_016533.5 | NP_057617.2 | 3 - 4 | 826 | 70 | |
GenBank mRNA | AF205633.1 | - | 3 - 4 | 501 | 70 |
BC057766.1 | - | 3 - 4 | 494 | 70 | |
BI597990.1 | - | 3 - 4 | 679 | 70 | |
BM128205.1 | - | 3 - 4 | 448 | 70 | |
CD695373.1 | - | 3 - 4 | 574 | 70 |